Learn More
CD2 Monoclonal Antibody (LO-CD2a), Functional Grade, In Vivo GOLD™, Leinco Technologies™

Rat Monoclonal Antibody
Supplier: Leinco Technologies Inc C509100MG


Description
Specificity: Anti-CD2 antibody (clone LO-CD2a) activity is directed against human CD2. Endotoxin Level: <1.0 EU/mg as determined by the LAL method. Purity: ≥95% monomer by analytical SEC and >95% by SDS Page. Preparation: Functional grade preclinical antibodies are manufactured in an animal free facility using in vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates. Storage: Functional grade preclinical antibodies may be stored sterile as received at 2-8C for up to one month. For longer term storage, aseptically aliquot in working volumes without diluting and store at -70C. Avoid Repeated Freeze Thaw Cycles.
CD2 Monoclonal antibody specifically detects CD2 in Human samples. It is validated for ELISA, Flow Cytometry, Functional AssaySpecifications
CD2 | |
Monoclonal | |
≥5.0 mg/mL | |
PBS with no preservative; pH 7.2-7.4 | |
P06729 | |
Rat | |
Protein A/G | |
RUO | |
914 | |
4°C short term, -80°C long term | |
Liquid |
ELISA, Flow Cytometry, Functional Assay | |
LO-CD2a | |
Functional Grade | |
CD2 | |
Cd2; CD2 antigen; CD2 antigen (p50), sheep red blood cell receptor; CD2 molecule; CD2R; cluster of differentiation 2; Erythrocyte receptor; FLJ46032; LFA2; LFA-2; LFA-3 receptor; Ly37; Ly-37; lymphocyte antigen 37; OX34; OX-34; OX-34 antigen; OX-45 surface antigen homolog to human T lymphocyte CD2 antigen; Rosette receptor; SRBC; surface immunoglobulin superfamily molecule; T11; T-cell surface antigen CD2; T-cell surface antigen T11/Leu-5 | |
Purified Recombinant Human CD2 | |
100 mg | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.