Learn More
CD2 Mouse anti-Human, Alexa Fluor™ 350, Clone: RPA-2.10, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP225200AF350
Catalog No. NP22520A350
Add to cart
Description
Description
CD2 Monoclonal antibody specifically detects CD2 in Human samples. It is validated for Western Blot,Flow Cytometry,Immunohistochemistry,Immunohistochemistry (Frozen).
Specifications
Specifications
CD2
RPA-2.10
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
CD2 antigen, CD2 antigen (p50), sheep red blood cell receptor, CD2 molecule, Erythrocyte receptor, FLJ46032, LFA-2, LFA-3 receptor, lymphocyte-function antigen-2, Rosette receptor, SRBC, T11, T-cell surface antigen CD2, T-cell surface antigen T11/Leu-5
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
Human
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
CD2
A recombinant protein was used as the immunogen for the CD2 antibody.
0.1 mL
Primary
914