Learn More
CD2 Mouse anti-Human, BV650, Clone: L303.1 (also known as L303), BD OptiBuild™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 745315
Catalog No. BDB745315
Add to cart
Description
Description
CD2 Monoclonal antibody specifically detects CD2 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD2
L303.1 (also known as L303)
Brilliant Violet 650
Mouse
50 μg
Refrigerate (2 to 8°C)
Monoclonal
Flow Cytometry
0.2 mg/mL
Purified
IgG2a, κ
RUO
Primary
Human