Learn More
CD2 Mouse anti-Human, FITC, Clone: 4AT2, Abnova™
Mouse monoclonal antibody raised against human CD2.
Manufacturer:  Abnova Corporation MAB12365
Catalog No. 89-933-758
Add to cart
Description
Description
Specifications
Specifications
CD2
4AT2
Mouse monoclonal antibody raised against human CD2.
In PBS, pH 7.4 (0.09% sodium azide, 0.2% BSA)
SRBC/T11
Mouse
IgG1
50 Reactions
Store at 4°C. Do not freeze.
Monoclonal
Human
Flow Cytometry, Immunofluorescence
FITC
Flow cytometry Immunofluorescence The optimal working dilution should be determined by the end user.
Liquid
CD2
CD2
Protein A/G affinity chromatography
RUO
Primary
914