Learn More
CD2 Mouse, anti-Human, R718, Clone: L303.1, BD OptiBuild™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 752112
Catalog No. BDB752112
Add to cart
Description
Description
The L303.1 monoclonal antibody specifically recognizes the human CD2 antigen. CD2 is a 45-50 kDa type I transmembrane glycoprotein that also forms the binding site for sheep erythrocytes. The CD2 antigen is present on approximately 75% of normal peripheral blood lymphocytes and 95% to 99% of thymocytes. It is also found on a subset of monocytes (approximately one-third) that might be precursors to dendritic cells. The CD2 antibody reacts with essentially all T lymphocytes and with a subset of NK lymphocytes. CD2 and CD58 have been shown to be coreceptors. The interaction of CD2 antigen and CD58 antigen facilitates antigen recognition by T lymphocytes.
Specifications
Specifications
CD2
L303.1
Liquid
50 μg
Refrigerate Upon Arrival
Monoclonal
Flow Cytometry
R718
Mouse
RUO
Primary
Human