Learn More
CD20 Human anti-Human, Alexa Fluor 700, Clone: Hu2, R&D Systems™
Human Monoclonal Antibody
Manufacturer:  R&D Systems FAB9575N100UG
Catalog No. FAB9575N100
Add to cart
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
CD20
Hu2
Flow Cytometry 0.25-1 ug/10^6 cells
Supernatant
Human
IgG1
100 ug
Primary
931
Human
Flow Cytometry
Alexa Fluor 700
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
B1, B-lymphocyte antigen CD20, B-lymphocyte cell-surface antigen B1, B-lymphocyte surface antigen B1, Bp35, Bp35MGC3969, CD20, CD20 antigen, CD20 receptor, CD20S7, CVID5, LEU-16, Leukocyte surface antigen Leu-16, Ly-44, Membrane-spanning 4-domains subfamily A member 1, membrane-spanning 4-domains, subfamily A, member 1, MS4A1, MS4A2, S7
Human CD20
Protein A or G purified from cell culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects Human CD20 based on Rituximab therapeutic antibody. This non-therapeutic antibody uses the same variable region sequence as the therapeutic antibody Rituximab. This product is for research use only.