Learn More
CD20, Mouse anti-Human,Feline, DyLight 350, Clone: B-Ly1, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP128380UV
Catalog No. NB004309
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
CD20 Monoclonal antibody specifically detects CD20 in Human, Feline samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).
Specifications
Specifications
CD20
B-Ly1
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Purified, Conjugated
MS4A1
The immunogen is a peptide made to the amino acid region CD20
Protein A or G purified
RUO
Store at 4°C in the dark.
Monoclonal
Human, Feline
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
B1, B-lymphocyte antigen CD20, B-lymphocyte cell-surface antigen B1, B-lymphocyte surface antigen B1, Bp35MGC3969, CD20 antigen, CD20 receptor, CD20S7, CVID5, LEU-16, Leukocyte surface antigen Leu-16, Membrane-spanning 4-domains subfamily A member 1, membrane-spanning 4-domains, subfamily A, member 1, MS4A2
Mouse
IgG1 Kappa
0.1 mL
Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer Stem Cells, Cell Biology, Cytokine Research, Immunology, Signal Transduction, Stem Cell Markers, Tumor Biomarkers
Primary
Human CD20