Learn More
CD20 Mouse anti-Human, Feline, FITC, Clone: B-Ly1, Southern Biotech™

Mouse Monoclonal Antibody
$90.30 - $229.00
Specifications
Antigen | CD20 |
---|---|
Clone | B-Ly1 |
Concentration | Lot specific |
Applications | Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
SouthernBiotech anti-human primary antibodies are available in a variety of conjugated formats including Alexa Fluor and Pacific Blue.
Specifications
CD20 | |
Lot specific | |
Monoclonal | |
Mouse | |
Feline, Human | |
IgG1 | |
Store at 2-8°C; Protect from light |
B-Ly1 | |
Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
FITC | |
RUO | |
Supplied in PBS/NaN3 | |
Primary |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.