Learn More
CD20 Mouse anti-Human, Primate, Alexa Fluor 405, Clone: 2H7, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP262206AF405
Catalog No. NB002397
Add to cart
Description
Description
CD20 Monoclonal specifically detects CD20 in Human, Primate samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Immunocytochemistry.
Specifications
Specifications
CD20
2H7
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Immunocytochemistry
Purified, Conjugated
MS4A1
Human tonsillar B cells
Protein A purified
Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer Stem Cells, Cell Biology, Cytokine Research, Immunology, Signal Transduction, Stem Cell Markers, Tumor Biomarkers
Primary
931
Human, Primate
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunocytochemistry
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
B1, B-lymphocyte antigen CD20, B-lymphocyte cell-surface antigen B1, B-lymphocyte surface antigen B1, Bp35MGC3969, CD20 antigen, CD20 receptor, CD20S7, CVID5, LEU-16, Leukocyte surface antigen Leu-16, Membrane-spanning 4-domains subfamily A member 1, membrane-spanning 4-domains, subfamily A, member 1, MS4A2
Mouse
IgG2b Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
The antibody MFG-06 reacts with a 40-45 kDa glycoprotein in human milk fat globule membrane preparation.