Learn More
CD20 Mouse, anti-Human, R718, Clone: 2H7, BD Horizon™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 566988
 View more versions of this product
Catalog No. BDB566988
Add to cart
Description
Description
The 2H7 monoclonal antibody specifically binds to CD20, encoded by the MS4A1 (Membrane-spanning 4-domains, subfamily A, member 1) gene. CD20 is a 33-37 kDa, unglycosylated four-transmembrane phosphoprotein. CD20 is expressed on pre-B-cells, resting and activated B cells, and follicular dendritic cells, but not plasma cells. Low level CD20 expression is observed on a small subset of normal circulating T lymphocytes. The CD20 molecule is involved in the regulation of B-cell activation.
Specifications
Specifications
CD20
2H7
Liquid
100 Tests
Refrigerate Upon Arrival
Monoclonal
Flow Cytometry
R718
Mouse
RUO
Primary
Human