Learn More
CD20 (Obinutuzumab) Recombinant Human Monoclonal Antibody (GA101), Functional Grade, Fc Muted™, Leinco Technologies™

Human Recombinant Monoclonal Antibody
Supplier: Leinco Technologies Inc LT912500UG


Description
Specificity: Obinutuzumab (GA101) activity is directed against human CD20. Endotoxin Level: <1.0 EU/mg as determined by the LAL method. Purity: ≥95% monomer by analytical SEC and >95% by SDS Page. Preparation: Functional grade preclinical antibodies are manufactured in an animal free facility using in vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates. Pathogen Testing: To protect mouse colonies from infection by pathogens and to assure that experimental preclinical data is not affected by such pathogens, all of Leinco recombinant biosimilar antibodies are tested and guaranteed to be negative for all pathogens in the IDEXX IMPACT I Mouse Profile. Storage: Functional grade biosimilar antibodies may be stored sterile as received at 2-8C for up to one month. For longer term storage, aseptically aliquot in working volumes without diluting and store at -80C. Avoid Repeated Freeze Thaw Cycles.
CD20 (Obinutuzumab) Recombinant Monoclonal antibody specifically detects CD20 (Obinutuzumab) in Human samples. It is validated for ELISA, Flow CytometrySpecifications
CD20 (Obinutuzumab) | |
Recombinant Monoclonal | |
≥5.0 mg/mL | |
PBS with no preservative; pH 7.2-7.4 | |
Human | |
Protein A/G | |
RUO | |
Human | |
Antibody | |
IgG1 κ |
ELISA, Flow Cytometry | |
GA101 | |
Functional Grade | |
afutuzumab; Gazyva; Gazyvaro; RG7159; RO5072759 | |
Humanized by grafting the complementarity-determining sequences of murine IgG1kappa antibody B-Ly1 onto human VH and VL acceptor frameworks | |
500 μg | |
Primary | |
4°C short term, -80°C long term | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.