Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD20 Rabbit anti-Human, Cynomolgus Monkey, Rhesus Macaque, Alexa Fluor™ 750, Clone: 10F381 (rituximab), Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP252646AF750

Catalog No. NP22646A70


Add to cart

Description

Description

CD20 Monoclonal antibody specifically detects CD20 in Human,Cynomolgus Monkey,Rhesus Macaque samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD20
10F381 (rituximab)
Flow Cytometry
B1, B-lymphocyte antigen CD20, B-lymphocyte cell-surface antigen B1, B-lymphocyte surface antigen B1, Bp35MGC3969, CD20 antigen, CD20 receptor, CD20S7, CVID5, LEU-16, Leukocyte surface antigen Leu-16, Membrane-spanning 4-domains subfamily A member 1, membrane-spanning 4-domains, subfamily A, member 1, MS4A2
Rabbit
IgG Kappa
Store at 4°C in the dark.
Monoclonal
CD20 (Human B-lymphocyte-restricted differentiation antigen, Bp35), is a hydrophobic transmembrane protein with a molecular weight of approximately 35 kDa located on pre-B and mature B lymphocytes, and most B-cell non-Hodgkin′s lymphomas, but is not found on stem cells, pro-B cells, normal plasma cells or other normal tissues .
Flow Cytometry
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
MS4A1
Human lymphoblastoid cell line SB.
0.1 mL
Primary
931
Cynomolgus Monkey, Human, Rhesus Macaque
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit