Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CD20, Rabbit Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry procedures with Thermo Scientific™ CD20, Rabbit Monoclonal Antibody.

Manufacturer:  Thermo Scientific™ RM2115S0

 View more versions of this product

Catalog No. RM2115S0


Add to cart

Description

Description

Obtain accurate, reproducible results in immunohistochemistry procedures with Thermo Scientific™ CD20, Rabbit Monoclonal Antibody, part of a comprehensive antibody portfolio. Studies show rabbit monoclonal antibodies are more sensitive and specific than other available markers.

CD20 is a non-Ig differentiation antigen of B cells and the expression of CD20 is restricted to normal and neoplastic B cells, being absent from all other leukocytes and tissues. It acts as calcium channel involved in B cell activation and cell cycle progression.

Host Species: Rabbit

Clone: EP459Y

Isotype: Rabbit IgG

Species Reactivity: Human. Others not tested.

Immunogen: A synthetic peptide corresponding the residues in the C-term of human CD20

Molecular Weight: 35kDa

Positive Control: Tonsil

Cellular Localization: Cell Membrane/cytoplasm

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

Immunohistochemistry (Paraffin)
EP459Y
Concentrated
A synthetic peptide corresponding the residues in the C-term of human CD20
100μL
Primary
Monoclonal
Monoclonal
Unconjugated
Rabbit
IgG
IVD
Primary
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit