Learn More
CD20, Rat anti-Human,Mouse, HRP, Clone: AISB12, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP143435H
Catalog No. NB004391
Add to cart
Description
Description
CD20 Monoclonal specifically detects CD20 in Human, Mouse samples. It is validated for Western Blot.
Specifications
Specifications
CD20
AISB12
Western Blot
Purified, Conjugated
MS4A1
The immunogen for this antibody was CD20.
Protein A or G purified
RUO
Store at 4°C in the dark.
Monoclonal
Western Blot
HRP
PBS with No Preservative
B1, B-lymphocyte antigen CD20, B-lymphocyte cell-surface antigen B1, B-lymphocyte surface antigen B1, Bp35MGC3969, CD20 antigen, CD20 receptor, CD20S7, CVID5, LEU-16, Leukocyte surface antigen Leu-16, Membrane-spanning 4-domains subfamily A member 1, membrane-spanning 4-domains, subfamily A, member 1, MS4A2
Rat
IgG2a
0.1 mL
Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer Stem Cells, Cell Biology, Cytokine Research, Immunology, Signal Transduction, Stem Cell Markers, Tumor Biomarkers
Primary
Human, Mouse