Learn More
CD200 Monoclonal Antibody (OX-90), Functional Grade, In Vivo PLATINUM™, Leinco Technologies™

Rat Monoclonal Antibody
Supplier: Leinco Technologies Inc C858100MG


Description
Specificity: Clone OX-90 reacts with murine CD200 (OX-2). Endotoxin Level: <0.5 EU/mg as determined by the LAL method. Purity: ≥98% monomer by analytical SEC and >95% by SDS Page. Preparation: Functional grade preclinical antibodies are manufactured in an animal free facility using in vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates. Pathogen Testing: To protect mouse colonies from infection by pathogens and to assure that experimental preclinical data is not affected by such pathogens, all of Leinco Purified Functional PLATINUM™ antibodies are tested and guaranteed to be negative for all pathogens in the IDEXX IMPACT I Mouse Profile. Storage: Functional grade preclinical antibodies may be stored sterile as received at 2-8C for up to one month. For longer term storage, aseptically aliquot in working volumes without diluting and store at -80C. Avoid Repeated Freeze Thaw Cycles.
CD200 Monoclonal antibody specifically detects CD200 in Mouse samples. It is validated for Flow Cytometry, Functional Assay, ImmunohistochemistrySpecifications
CD200 | |
Monoclonal | |
≥5.0 mg/mL | |
PBS with no preservative; pH 7.2-7.4 | |
O54901 | |
Rat | |
Protein A/G | |
RUO | |
17470 | |
4°C short term, -80°C long term | |
Liquid |
Flow Cytometry, Functional Assay, Immunohistochemistry | |
OX-90 | |
Functional Grade | |
Cd200 | |
antigen identified by monoclonal antibody MRC OX-2; CD200; Cd200 antigen; Cd200 molecule; cell surface protein (thymocyte antigen identified by monoclonal antibody MRC-OX2); Cspmo2; hypothetical protein LOC534910; MOX1; MOX2; MRC; MRC OX-2 antigen; MRCOX2; My033; OX2; OX-2; OX-2 membrane glycoprotein | |
Mouse CD200-rat CD4 fusion protein | |
100 mg | |
Primary | |
Mouse | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.