Learn More
CD200 Mouse anti-Human, BB515, Clone: MRC OX-104, BD Horizon™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 566024
Catalog No. BDB566024
Add to cart
Description
Description
CD200 Monoclonal antibody specifically detects CD200 in Human samples. It is validated for Flow cytometry.
Specifications
Specifications
CD200
MRC OX-104
Vol. Per test: 5μL
Purified
Mouse
Mouse IgG1, κ
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB515 under optimum conditions and unconjugated antibody was removed.
Monoclonal
Flow Cytometry
BB515
Aqueous buffered solution containing ≤0.09% sodium azide.
OX-2; MOX1; MOX2 ; My033
Human CD200 Recombinant Protein
25 Tests
Cell Biology
Primary
Human
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Reseach Use Only.