Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ CD200/OX2 Antibody Pair [HRP]
Polyclonal/Monoclonal Antibody Pair
$832.00
Specifications
Assay Range | 46.88 pg/mL |
---|---|
Assay Sensitivity | 46.88-3000 pg/mL |
Assay | Sandwich ELISA |
Conjugate | HRP |
Product Type | Antibody Pairs |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB009665
|
Novus Biologicals™
NBP27931315PLATES |
15 Plates |
Each of 1 for $832.00
|
|
|||||
Description
This solid phase sandwich ELISA utilizes an antibody specific for Human CD200/OX2.Specifications
46.88 pg/mL | |
Sandwich ELISA | |
Antibody Pairs | |
Human | |
antigen identified by monoclonal MRC OX-2, CD200 antigenMOX1, CD200 molecule, MOX2MRC, MRC OX-2 antigen, OX-2, OX-2 membrane glycoprotein | |
CD200 | |
Storage is content dependent. | |
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 3000 pg/mL is recommended. |
46.88-3000 pg/mL | |
HRP | |
This solid phase sandwich ELISA utilizes an antibody specific for Human CD200/OX2. | |
Sandwich ELISA | |
4345 | |
Rabbit | |
RUO | |
CD200/OX2 |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title