Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD200 Rat anti-Mouse , BV421, Clone: OX-90 (also known as OX90), BD Biosciences™
Rat Monoclonal Antibody
Supplier: BD Biosciences 565547
Description
CD200 Monoclonal antibody specifically detects CD200 in Mouse samples. It is validated for Flow CytometrySpecifications
CD200 | |
Monoclonal | |
Brilliant Violet 421 | |
Cd200; OX2; OX-2; OX2G; MRC OX-2 antigen; Mox2 | |
Soluble fusion protein of the extracellular region of mouse OX-2 antigen w/domains 3 & 4 of rat CD4 | |
RUO | |
Mouse | |
Rat IgG2a κ |
Flow Cytometry | |
OX-90 (also known as OX90) | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Rat | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BV421 under optimum conditions, and unconjugated antibody and free BD Horizon BV421 were removed. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title