Learn More
Abnova™ CD200R1 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00131450T01
Catalog No. 89-938-624
Add to cart
Description
Description
Specifications
Specifications
Western Blot
CD200R1 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 38.39kDa
Transient overexpression cell lysate was tested with anti-CD200R1 antibody (Mfr. No. H00131450-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
293T
NM_138806.3
pCMV-CD200R1 full-length
100 μL
Human