Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD200R1, anti-Human, Clone: OX108, Abnova™

Monoclonal Antibody

Manufacturer:  Abnova Corporation RAB00113

Catalog No. 89-032-456


Add to cart

Specifications

Specifications

CD200R1
OX108
Flow Cytometry (1 ug/mL) The optimal working dilution should be determined by the end user.
CD200 receptor 1
CD200R|HCRTR2|MOX2R|OX2R
Original antibody is raised against recombinant protein hCD200RCD4d3+4.
200 μg
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Primary
131450
Flow Cytometry
Unconjugated
In PBS, pH 7.2 (0.02% Proclin 300).
Liquid
CD200R1
IgG, kappa
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit