Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD200R1 Antibody (OX-110), Biotin, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NB10063562B
Description
CD200R1 Monoclonal antibody specifically detects CD200R1 in Mouse samples. It is validated for Flow CytometrySpecifications
CD200R1 | |
Monoclonal | |
Biotin | |
CD200 receptor 1, CD200RMOX2Rcell surface glycoprotein CD200 receptor 1, Cell surface glycoprotein OX2 receptor 1, cell surface glycoprotein receptor CD200, CRTR2, HCRTR2, MOX2 receptor, OX2RCD200 cell surface glycoprotein receptor | |
Fusion protein mCD200RCD4d3+4 | |
0.125 mL | |
Cancer, Neuronal Cell Markers, Neuroscience | |
131450 | |
Store at 4°C in the dark. | |
IgG2a |
Flow Cytometry | |
OX-110 | |
PBS | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction