Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD200R1 Mouse anti-Human, Janelia Fluor 549, Clone: OX108, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP252694JF549

Catalog No. NBP52694549


Add to cart

Description

Description

CD200R1 Monoclonal antibody specifically detects CD200R1 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD200R1
OX108
Flow Cytometry
Purified
CD200R1
Recombinant protein - hCD200RCD4d3+4.
Protein A or G purified
Cancer, GPCR, Neuronal Cell Markers, Neuroscience
Primary
131450
Human
Flow Cytometry
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
CD200 receptor 1, CD200RMOX2Rcell surface glycoprotein CD200 receptor 1, Cell surface glycoprotein OX2 receptor 1, cell surface glycoprotein receptor CD200, CRTR2, HCRTR2, MOX2 receptor, OX2RCD200 cell surface glycoprotein receptor
Mouse
IgG1 Kappa
0.1mL
Store at 4°C in the dark.
Monoclonal
Recognizes human CD200R, a receptor for the CD200/OX2.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit