Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD200R1L Antibody (2692A), PE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB10198P
This item is not returnable.
View return policy
Description
CD200R1L Monoclonal antibody specifically detects CD200R1L in Mouse samples. It is validated for Flow CytometrySpecifications
CD200R1L | |
Monoclonal | |
PE | |
cell surface glycoprotein CD200 receptor 2, CD200 receptor 1-like, CD200R2, CD200RLa | |
Mouse myeloma cell line NS0-derived mouse CD200R1L, Ile25-Thr220, Accession # Q6XJV6.1 | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Flow Cytometry | |
2692A | |
PBS | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
344807 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction