Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ CD200R1L Antibody Pair [HRP]

Monoclonal Antibody Pair

Manufacturer:  Novus Biologicals™ NBP2796615PLATES

 View more versions of this product

Catalog No. NB010362


Add to cart

Description

Description

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human CD200R1L.
Specifications

Specifications

Sandwich ELISA
Antibody Pairs
125 pg/mL
344807
RUO
CD200R1L
5 Plates
125-8000 pg/mL
cell surface glycoprotein CD200 receptor 2, CD200 receptor 1-like, CD200R2, CD200RLa
Conjugated
HRP
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human CD200R1L.
Sandwich ELISA
  • Detection Antibody: 0.25 mg/mL mouse anti-CD200R1L monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % glycerol, pH 7.4).
  • Capture Antibody: 0.5 mg/mL of rabbit anti-CD200R1L monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 80 ng of recombinant CD200R1L.
  • Storage is content dependent.
    Mouse/Rabbit
    Human
    CD200R1L
    1. Capture Antibody: Dilute to a working concentration of 0.5 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.25 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 8 ng/mL is recommended.
    SDS
    Documents
    Product Certifications
    Promotions

    Promotions

    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit