Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD202b (TIE2) Rat anti-Mouse, Biotin, Clone: TEK4, eBioscience™ Non-distribution product as customer accommodation.

Rat Monoclonal Antibody

Manufacturer:  Invitrogen 13598785

 View more versions of this product

Catalog No. 50-121-01


Add to cart

Description

Description

Description: The TEK4 monoclonal antibody reacts with mouse Tie-2, also known as CD202. A member of the tyrosine kinase receptor family, Tie-2 is expressed on endothelial and a subset of hematopoietic cells and is believed to play a role in both angiogenesis and hematopoiesis during development of the mouse embryo. In fetal liver and adult bone marrow, Tie-2 is expressed by a subpopulation of hematopoietic progenitor cells characterized as Lineage markers^-, c-Kit^+, Sca1^+ cells. Long-term multilineage repopulating cells were detected in Tie-2^+, Lineage^-, c-Kit^+, Sca1^+ cells but not in Tie-2^-, Lineage^-, c-Kit^+, Sca1^+ cells. Applications Reported: The TEK4 antibody has been reported for use in flow cytometric analysis. Applications Tested: This TEK4 antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 0.5 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filtration: 0.2 µm post-manufacturing filtered.

The TEK receptor tyrosine kinase is expressed almost exclusively in endothelial cells in mice, rats, and humans. This receptor possesses a unique extracellular domain containing 2 immunoglobulin-like loops separated by 3 epidermal growth factor-like repeats that are connected to 3 fibronectin type III-like repeats. The ligand for the receptor is angiopoietin-1. Defects in TEK are associated with inherited venous malformations; the TEK signaling pathway appears to be critical for endothelial cell-smooth muscle cell communication in venous morphogenesis. TEK is closely related to the TIE receptor tyrosine kinase.
Specifications

Specifications

CD202b (TIE2)
Monoclonal
0.5 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q02858
CD202B, TIE-2, TIE2, VMCM, VMCM1, angiopoietin-1 receptor, endothelial tyrosine kinase, hTIE2, p140 TEK, soluble TIE2 variant 1, soluble TIE2 variant 2, tunica interna endothelial cell kinase, tyrosine kinase with Ig and EGF homology domains-2, tyrosine-protein kinase receptor TEK, tyrosine-protei
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
TEK4
Biotin
Tek
Liquid
Tek
IgG1, kappa
500 μg
4° C, store in dark, DO NOT FREEZE!
Primary
21687
SDS
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.