Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD204 Mouse anti-Human, PE-Cyanine7, Clone: PSL204, eBioscience™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

$169.00 - $349.00

Specifications

Antigen CD204
Clone PSL204
Target Species Human
Conjugate PE-Cyanine7
Isotype IgG1, kappa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
25204541 Non-distribution product as customer accommodation. View Documents Invitrogen
25204541
25 Tests Each for $169.00
Add to cart
 
25204542 Non-distribution product as customer accommodation. View Documents Invitrogen
25204542
100 Tests Each for $349.00
Add to cart
 
Description

Description

This monoclonal antibody PSL204 reacts with human CD204, also known as Macrophage Scavenger Receptor 1 (MSR1), Scavenger Receptor Type A (SR-A), or SCARA1. It is expressed by multiple populations of macrophages and dendritic cells. CD204 belongs to the class A family of scavenger receptors, a group of five homotrimetric transmembrane proteins that contain a short cytoplasmic tail and extracellular C -terminus. Other members of this family include: MARCO (SCARA2), CSR (SCARA3), SRCL (SCARA4) and SCARA5. CD204 binds and internalizes oxidized low density lipoproteins (LDL), which drives macrophage differentiation into foam cells, secretion of inflammatory cytokines, and development of atherosclerotic lesions. In addition to oxidized LDL, CD204 binds and uptakes a wide variety of macromolecules, including bacterial cell wall components, viral DNA, calciprotein complexes, apoptotic cells, and beta-amyloid fibrils. This broad binding specificity implicates CD204-expressing macrophages in a number of physiological processes and pathologies, such as host defense, inflammation, bone remodeling, atherosclerosis, sepsis, and Alzheimer ft.s disease. In humans, CD204 exists as 3 splice variants. The more ubiquitous full length isoform (type I, SR-AI), a shorter isoform (type II, SR-AII) that lacks the C-terminal cysteine-rich domain, and the shortest isoform (type III, SR-AIII).

Isoforms I and II bind modified LDL with similar affinity, whereas isoform III is not expressed on the cell surface. This monoclonal antibody PSL204 binds only the long (type I, SR-AI) isoform. The PSL204 will recognize CD204 on cells that have been formaldehyde-fixed and permeabilized. This antibody does not cross-react with mouse or rat CD204. This PSL204 antibody has been pre-titrated and tested by flow cytometric analysis of human monocyte-derived macrophages. This can be used at 5 µL (0.06 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 µL of cell sample + 100 µL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Filtration: 0.2 µm post-manufacturing filtered. CD204 gene encodes the class A macrophage scavenger receptors, which include three different types (1, 2, 3) generated by alternative splicing of this gene. These receptors or isoforms are macrophage-specific trimeric integral membrane glycoproteins and have been implicated in many macrophage-associated physiological and pathological processes including atherosclerosis, Alzheimer ft.s disease, and host defense. The isoforms type 1 and type 2 are functional receptors and are able to mediate the endocytosis of modified low density lipoproteins (LDLs). The isoform type 3 does not internalize modified LDL (acetyl-LDL) despite having the domain shown to mediate this function in the types 1 and 2 isoforms. It has an altered intracellular processing and is trapped within the endoplasmic reticulum, making it unable to perform endocytosis. The isoform type 3 can inhibit the function of isoforms type 1 and type 2 when co-expressed, indicating a domit negative effect and suggesting a mechanism for regulation of scavenger receptor activity in macrophages. Diseases associated with MSR1 include Barrett Esophagus and Prostate Cancer.
Specifications

Specifications

CD204
Human
IgG1, kappa
Monoclonal
Liquid
Antibody
4° C, store in dark, DO NOT FREEZE!
PSL204
PE-Cyanine7
Flow Cytometry
Primary
Monoclonal
PBS with 0.2% BSA and 0.09% sodium azide
RUO

For Research Use Only

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit