Learn More
CD205 Mouse anti-Human, Clone: HD30, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA528126
Catalog No. PIMA528126
Add to cart
Description
Description
CD205 (LY25, Lymphocyte Antigen 75) is an integral membrane glycoprotein involved in antigen uptake, trafficking and presentation that improves the induction of antigen-specific T cell immunity. CD205 is highly expressed by CD8+ dendritic cells (DCs) and also expressed at different levels by bone marrow Gr1+ cells, Langerhans cells, (BMDC) bone marrow derived DCs and thymic epithelial cells. CD205 acts as an endocytic receptor that directs captured antigens to the extracellular space towards a specialized antigen processing region. Further, CD205 also prompts the reduced proliferation of B lymphocytes. Diseases associated with CD205 dysfunction include pneumonic plague and lipid pneumonia.
Specifications
Specifications
CD205
Monoclonal
1 mg/mL
PBS with no preservative
O60449
LY75
Purified extracellular domain of human CD205-IgG1 Fc fusion protein
Protein G
RUO
Antibody
Monoclonal
Human
Flow Cytometry, Immunohistochemistry (Frozen), Western Blot
HD30
Unconjugated
LY75
Liquid
Mouse
IgG1
500 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
4065