Learn More
CD21 Mouse anti-Baboon, Human, Porcine, Rhesus, Brilliant Ultraviolet 737, Clone: B-ly4, BD Horizon™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 612788
Catalog No. BDB612788
Description
Description
CD21 Monoclonal antibody specifically detects CD21 in Baboon,Human,Porcine,Rhesus samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD21
B-ly4
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse
Affinity Chromatography
RUO
Primary
Baboon, Human, Porcine, Rhesus Monkey
Flow Cytometry
Brilliant Ultraviolet 737
Affinity purified
IgG1, kappa
100 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal