Learn More
CD21 Mouse anti-Human, Alexa Fluor 594, Clone: 544408, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB4909T100UG
Catalog No. FAB4909T100
Add to cart
This item is not returnable. View return policy
Description
Description
CD21 Monoclonal specifically detects CD21 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD21
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
P20023
CR2
Chinese hamster ovary cell line CHO-derived recombinant human CD21 Ile21-Arg971 Accession # P20023
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects human CD21 in direct ELISAs and Western blots.
Flow Cytometry
544408
Alexa Fluor 594
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
C3DR, C3DRSLEB9, CD21, CD21 antigen, Complement C3d receptor, complement component (3d/Epstein Barr virus) receptor 2, complement receptor type 2, Cr2, EBV receptor, Epstein-Barr virus receptor
Mouse
IgG1
RUO
Primary
1380
Human
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)