Learn More
CD21 Mouse anti-Human,Baboon,Cynomolgus,Pig,Rhesus Ma, BB700, Clone: B-ly4, BD Horizon™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 566570
 View more versions of this product
Catalog No. BDB566570
Add to cart
Description
Description
CD21 Monoclonal antibody specifically detects CD21 in Human,Baboon,Cynomolgus,Pig,Rhesus Ma samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD21
B-ly4
Purified
IgG1, κ
RUO
Primary
Human, Baboon, Cynomolgus, Pig, Rhesus Macaque
Flow Cytometry
BB700
Mouse
25 Tests
Refrigerate (2 to 8°C)
Monoclonal