Learn More
CD21 Mouse anti-Human, Brilliant Ultraviolet 737, Clone: B-ly4, BD Horizon™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 612789
Catalog No. BDB612789
Add to cart
Description
Description
CD21 Monoclonal antibody specifically detects CD21 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD21
B-ly4
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse
Affinity Chromatography
RUO
Primary
Human
Flow Cytometry
Brilliant Ultraviolet 737
Affinity purified
IgG1, kappa
25 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal