Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD21 Mouse anti-Human, Clone: 1F8, Novus Biologicals™
Mouse Monoclonal Antibody
$177.03 - $436.00
Specifications
Antigen | CD21 |
---|---|
Clone | 1F8 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
CD21 Monoclonal specifically detects CD21 in Human, Monkey samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CD21 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Dendritic Cell Markers, Immunology | |
C3DRSLEB9, CD21, CD21 antigen, Complement C3d receptor, complement component (3d/Epstein Barr virus) receptor 2, complement receptor type 2, Cr2, EBV receptor, Epstein-Barr virus receptor | |
CR2 | |
IgG1 κ | |
Tissue culture supernatant |
1F8 | |
Monoclonal | |
Supernatant | |
RUO | |
P20023 | |
1380 | |
Purified receptor for the C3d fragment of the third complement component CR2, from human tonsil. | |
Primary | |
Store at 4°C. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title