Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD21 Monoclonal Antibody (HB5), NovaFluor™ Yellow 660, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Specifications
Antigen | CD21 |
---|---|
Clone | HB5 |
Concentration | 0.05 μg/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
H055T02Y04
![]() |
LIFE TECHNOLOGIES
H055T02Y04 |
25 Tests | This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
H055T03Y04
![]() |
LIFE TECHNOLOGIES
H055T03Y04 |
100 Tests | This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
CD21 Monoclonal antibody specifically detects CD21 in Human samples. It is validated for Flow CytometrySpecifications
CD21 | |
0.05 μg/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
C3b/C4b receptor; C3-binding protein; C3BR; C3DR; C4BR; CD21; CD35; CD35 antigen; cell surface receptor for C3d; complement C3b/C4b receptor 1 (Knops blood group); complement C3d receptor; Complement C3d receptor (C3DR); complement C3d receptor 2; complement component (3b/4b) receptor 1 (Knops blood group); complement component (3d/Epstein Barr virus) receptor 2; complement component 3b/4b receptor 1 (Knops blood group); complement component 3d receptor 2; complement component receptor 2; complement receptor 1 long isoform; complement receptor 2; complement receptor type 1; complement receptor type 1-like protein; complement receptor type 2; Complement Receptor type 2 (CR2); CR; CR1; Cr-1; CR1-L; Cr2; Cr-2; CVID7; EBV receptor; EBV-R; Epstein-Barr virus receptor; EVBR; expressed in B lymphocytes; KN; Knops blood group antigen; LOW QUALITY PROTEIN: complement receptor type 1; SLEB9 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
HB5 | |
Flow Cytometry | |
NovaFluor Yellow 660 | |
Mouse | |
Human | |
P20023 | |
1380 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
CR2 |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title