Learn More
CD217 (IL-17RA) Mouse anti-Human, BB700, Clone: 133617, BD OptiBuild™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 747946
Catalog No. BDB747946
Add to cart
Description
Description
CD217 (IL-17RA) Monoclonal antibody specifically detects CD217 (IL-17RA) in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD217 (IL-17RA)
133617
BB700
Mouse
50 μg
Refrigerate (2 to 8°C)
Monoclonal
Human
Flow Cytometry
0.2 mg/mL
Conjugated, Purified
IgG1, κ
RUO
Primary
CD217 (IL-17RA)