Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ CD22 Ab-4, Mouse Monoclonal Antibody

Obtain accurate and reproducible results in immunohistochemistry experiments with Thermo Scientific™ CD22 Ab-4, Mouse Monoclonal Antibody.

Specifications

Antigen CD22 Ab-4
Clone FPC1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MS1087R1
View Documents
Thermo Scientific™
MS1087R1
1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Obtain accurate and reproducible results in immunohistochemistry experiments with Thermo Scientific™ CD22 Ab-4, Mouse Monoclonal Antibody, one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.

CD22 expression is restricted to normal and neoplastic B cells and is absent from other haemopoietic cell types.In B-cell ontogeny, CD22 is first expressed in the cytoplasm of pro-B and pre-B cells, and on the surface as B cells mature to become IgD+. It is not expressed by plasma cells. CD22 is found highly expressed in follicular mantle and marginal zone B-cells, while germinal center B-cells are relatively weak. CD22 is a member of the immunoglobulin superfamily and serves as an adhesion receptor for sialic acid-bearing ligands expressed on erythrocytes and all leukocyte classes. It also associates with tyrosine kinases and play a role in signal transduction and B-cell activation.

Host Species: Mouse

Clone: FPC1

Isotype: IgG1

Species Reactivity: Human. Others not tested.

Epitope: Extracellular

Immunogen: Recombinant protein encoding the external domain of human CD22

Molecular Weight: 130-140kDa

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin only)

Specifications

Specifications

CD22 Ab-4
Immunohistochemistry (Paraffin)
Unconjugated
IVD
Primary
FPC1
Monoclonal
Murine
B-Cell
Recombinant Protein encoding the external domain of human CD22
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit