Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD223 (LAG-3) Rat anti-Mouse, Clone: eBioC9B7W (C9B7W), eBioscience™ Non-distribution product as customer accommodation.

Rat Monoclonal Antibody

Manufacturer:  Invitrogen 14223181

 View more versions of this product

Catalog No. 50-129-94


Add to cart

Description

Description

Description: The eBioC9B7W monoclonal antibody recognizes mouse CD223 (LAG-3, LAG3) protein expressed by activated alpha/beta-TCR bearing T cells, a subset of gamma/delta-TCR bearing T cells and a subset of NK cells. CD223 is a 70 kDa type I transmembrane protein with a structure that is similar to CD4. However, a soluble form of human CD223 has been detected by ELISA in human serum, and data suggest that mouse CD223 is proteolytically cleaved in the D4 domain. This results in a 54 kDa fragment containing all the extracellular domains, and a 16 kDa fragment containing the intracellular and transmembrane domains. The 54 kDa can remain membrane-associated or be released as soluble CD223. CD223 binds to MHC class II with higher affinity than CD4, and it is thought that this interaction is involved in the negative regulation of T-cell activation and homeostatic proliferation. Furthermore, CD223 is expressed by CD4+CD25+ regulatory T cells, and it has been suggested that CD223 may be involved in their regulatory function. Applications Reported: This eBioC9B7W (C9B7W) antibody has been reported for use in flow cytometric analysis, immunoprecipitation, and ELISA. (Please use Functional Grade purified eBioC9B7W (C9B7W), cat. 16-2231, in functional assays.). Applications Tested: This eBioC9B7W (C9B7W) antibody has been tested by flow cytometric analysis of anti-CD3 and anti-CD28-activated mouse splenocytes. This can be used at less than or equal to 0.5 µg per test.

A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. This eBioC9B7W (C9B7W) antibody has also been tested by ELISA analysis of supernatant produced from mouse CD223-transfected 3A9 cells. This can be used in ELISA at 2 µg/mL for capture of the secreted form of CD223, together with the polyclonal anti-mouse CD223 (cat. 24-2232) as the detection antibody. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 µm post-manufacturing filtered. LAG-3 is a 70-kDa surface glycoprotein belonging to the Ig superfamily with homology to CD4. LAG-3 binds to MHC class II with higher affinity than CD4 and is thought to be involved in the negative regulation of T cell activation and homeostatic proliferation. Surface expression of LAG-3 has been reported on activated T cells (including regulatory T cells) and NK cells. CD8+ T cells usually express LAG-3 at significantly higher levels than CD4+ T cells. Coexpression of LAG-3 and CD49b has been proposed to identify human and mouse Type 1 regulatory T cells (Tr1 cells).
Specifications

Specifications

CD223 (Lag-3)
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Q61790
CD223, LAG-3, Ly66
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
ELISA, Flow Cytometry, Functional Assay, Immunoprecipitation
eBioC9B7W (C9B7W)
Unconjugated
Lag3
Liquid
Lag3
IgG1, kappa
50 μg
4° C
Primary
16768
SDS
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.