Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CD223 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA582490
Description
Immunogen sequence: LVTGDNGDFT LRLEDVSQAQ AGTYTCHIHL QEQQLNATVT LAIITVTPKS FGSPGSLGKL LCEVTPVSGQ ERFVWSSLDT PSQRSFSGPW LEAQEAQLLS QPWQCQLYQG ERLLGAAVYF TELSSPGAQ.
LAG-3 is a 70-kDa surface glycoprotein belonging to the Ig superfamily with homology to CD4. LAG-3 binds to MHC class II with higher affinity than CD4 and is thought to be involved in the negative regulation of T cell activation and homeostatic proliferation. Surface expression of LAG-3 has been reported on activated T cells (including regulatory T cells) and NK cells. CD8+ T cells usually express LAG-3 at significantly higher levels than CD4+ T cells. Coexpression of LAG-3 and CD49b has been proposed to identify human and mouse Type 1 regulatory T cells (Tr1 cells).
Specifications
CD223 | |
Polyclonal | |
Unconjugated | |
LAG3 | |
Activation-induced cytidine deaminase-linked autoimmunity protein; Aida; CD223; FDC; LAG3; LAG-3; Ly66; lymphocyte activating 3; lymphocyte activation gene 3 protein; lymphocyte-activation gene 3; Secreted lymphocyte activation gene 3 protein; sLAG 3; sLAG3; sLAG-3; soluble LAG 3lymphocyte activating 3; soluble LAG3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3902 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P18627 | |
LAG3 | |
Recombinant protein corresponding to Human LAG3. Recombinant protein control fragment (Product #RP-90012). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction