Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD229/SLAMF3/Lymphocyte Antigen 9 Mouse anti-Human, Allophycocyanin/Cy7, Clone: MM0163-3L25, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP212151APCCY7

Catalog No. N212151APC7


Add to cart

Description

Description

CD229/SLAMF3/Lymphocyte Antigen 9 Monoclonal antibody specifically detects CD229/SLAMF3/Lymphocyte Antigen 9 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD229/SLAMF3/Lymphocyte Antigen 9
MM0163-3L25
Flow Cytometry
Conjugated, Purified
LY9
Human Lymphocyte Antigen 9
Protein G purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
CD229, CD229 antigen, cell-surface molecule Ly-9, hly9, lymphocyte antigen 9Cell surface molecule Ly-9, mLY9, SLAMF3, T-lymphocyte surface antigen Ly-9
Mouse
IgG1
0.1 mL
Primary
4063.0
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit