Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD229/SLAMF3/Lymphocyte Antigen 9 Antibody (HLy9.1.25) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP1424220.05MG
Description
CD229/SLAMF3/Lymphocyte Antigen 9 Monoclonal specifically detects CD229/SLAMF3/Lymphocyte Antigen 9 in Human, Mouse samples. It is validated for Flow Cytometry, Immunoprecipitation, Functional, CyTOF-ready.Specifications
CD229/SLAMF3/Lymphocyte Antigen 9 | |
Monoclonal | |
1.0 mg/mL | |
Western Blot, Flow Cytometry 1:10-1:1000, Immunoprecipitation 1:10-1:500, Functional, CyTOF-ready | |
CD229, CD229 antigen, cell-surface molecule Ly-9, hly9, lymphocyte antigen 9Cell surface molecule Ly-9, mLY9, SLAMF3, T-lymphocyte surface antigen Ly-9 | |
Mouse | |
0.05 mg | |
4063 | |
Human, Mouse | |
IgG1 |
Western Blot, Flow Cytometry, Immunoprecipitation, Functional Assay, CyTOF | |
HLy9.1.25 | |
Unconjugated | |
PBS with 0.09% sodium azide | |
LY9 | |
Mouse pre-B cell line 300.19 transfected with CD229 cDNA | |
Primary | |
Recognizes the human CD229 cell surface antigen, a 100-120kD cell surface glycoprotein that is the homologue of murine Ly-9. CD229 is a member of the immunoglobulin superfamily and may have a function in cell signalling. The antigen is expressed at low levels by mature T and B lymphocytes and at higher levels by single-positive thymocytes. Expression at variable levels is also seen in some cell lines, such as Daudi, U937 and HUT78. 19 transfected with CD229 cDNA | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction