Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD23 Mouse anti-Human, BB515, Clone: M-L233, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 564555
Description
CD23 Monoclonal antibody specifically detects CD23 in Human samples. It is validated for Flow Cytometry.Specifications
CD23 | |
Monoclonal | |
Brilliant Blue 515 (BB515) | |
100 Tests | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG1 κ |
Flow Cytometry | |
M-L233 | |
Mouse | |
RUO | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction