Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD23 Mouse anti-Human, Unconjugated, Clone: EBVCS1, MilliporeSigma™
Mouse Monoclonal Antibody
$184.00 - $463.00
Specifications
Antigen | CD23 |
---|---|
Clone | EBVCS1 |
Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Radioimmune Assays (RIA), Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABF23172MI
|
MilliporeSigma
MABF231725UG |
25 μg |
Each for $184.00
|
|
|||||
MABF23171MI
|
MilliporeSigma
MABF2317100UG |
100 μg |
Each for $463.00
|
|
|||||
Description
CD23 Monoclonal antibody specifically detects CD23 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Radioimmunoassay, Western Blotting.Specifications
CD23 | |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Radioimmune Assays (RIA), Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Low affinity immunoglobulin epsilon Fc receptor;BLAST-2;C-type lectin domain family 4-member J;Fc-epsilon-RII;Immunoglobulin E-binding factor;Lymphocyte IgE receptor;CD23 | |
Epstein-Barr Virus transformed human cells. | |
Primary | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
∼45 kDa observed; 36.47 kDa calculated. Uncharacterized bands may be observed in some lysate(s). |
EBVCS1 | |
Monoclonal | |
Purified | |
Inflammation and Immunology | |
NP_001193948 | |
FCER2, CD23A, CLEC4J, FCE2, IGEBF | |
IgG1 | |
Protein G Purified | |
Clone EBVCS1 is a mouse monoclonal antibody that specifically detects CD23 on Epstein-Barr virus transformed cells. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title