Learn More
CD231 (TALLA-1) Mouse anti-Human, PE, Clone: M3-3D9 (also known as SN1a), BD Pharmingen™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 566578
Catalog No. BDB566578
Add to cart
Description
Description
CD231 (TALLA-1) Monoclonal antibody specifically detects CD231 (TALLA-1) in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD231 (TALLA-1)
M3-3D9 (also known as SN1a)
PE
Mouse
50 μg
Refrigerate (2 to 8°C)
Monoclonal
Flow Cytometry
0.2 mg/mL
Purified
IgG1, κ
RUO
Primary
Human