Learn More
Epredia™ Lab Vision™ CD24 (GPI-Linked Surface Mucin) Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide
Ensure accurate, reproducible results in immunohistochemistry, western blotting, and flow cytometry experiments with Epredia™ CD24 (GPI-Linked Surface Mucin) Ab-1.
Supplier: Epredia™ MS1278P1ABX
Description
CD24 is a two-chain glycosylphosphatidylinositol (GPI)-anchored glycoprotein expressed at multiple stages of B-cell development, beginning with the bone marrow pro-B-cell compartment and continuing through mature, surface Ig positive B-cells. Plasma cell expression is very low or negative. It is also expressed on the majority of B-lineage acute lymphoblastic leukemias, B-cell CCLs and B-cell non-Hodgkin's lymphomas. CD24 may play a role in regulation of B-cell proliferation and maturation.Host Species: Mouse
Clone: SN3
Isotype: IgG1/κ
Species Reactivity: Human. Others not tested.
Epitope: Not determined
Immunogen: Cell membrane antigen preparation isolated from NALM-1 human pre-B leukemia cell line
Molecular Weight: 30-70 kDa
Positive Control: Tonsil
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Frozen only)
- Western Blotting
- Flow Cytometry
Note:
USA: RUO; Int'l: RUO
Specifications
CD24 (GPI-Linked Surface Mucin) Ab-1 | |
Monoclonal | |
1mg/mL | |
Mouse | |
100 μL | |
B-Cell | |
Human | |
IgG2a κ |
Flow Cytometry, Immunohistochemistry (Frozen), Western Blot | |
SN3 | |
Unconjugated | |
Cell membrane antigen preparation isolated from NALM-1 human pre-B leukemia cell line | |
RUO | |
Primary | |
Concentrated |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.