Learn More
CD24 Mouse anti-Human, Alexa Fluor 700, Clone: 4F4E10, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237352AF700
Catalog No. NBP237352X
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
CD24
4F4E10
Western Blot, Flow Cytometry, ELISA
Purified
CD24
Purified recombinant fragment of human CD24 (AA: 15-80) expressed in E. Coli.
Protein A or G purified
B Cell Development and Differentiation Markers, Cancer, Neuronal Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers
Primary
100133941
Western Blot, Flow Cytometry, ELISA
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
CD 24, CD24 antigen, CD24 antigen (small cell lung carcinoma cluster 4 antigen), CD24 molecule, CD24Asignal transducer CD24, FLJ22950, FLJ43543, MGC75043, Small cell lung carcinoma cluster 4 antigen
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human