Learn More
CD24 Mouse anti-Human, BB515, Clone: ML5, BD Horizon™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 564522
Catalog No. BDB564522
Add to cart
Description
Description
CD24 Monoclonal antibody specifically detects CD24 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD24
ML5
Purified
IgG2a, κ
RUO
Primary
Human
Flow Cytometry
BB515
Mouse
25 Tests
Refrigerate (2 to 8°C)
Monoclonal