Learn More
CD24 Mouse anti-Human, FITC, Clone: SN3, Invitrogen™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MHCD2401
Catalog No. 50-112-1214
Add to cart
Description
Description
Commonly used, FITC conjugates provide relatively high absorptivity, excellent fluorescence quantum yield, and good water solubility.
CD24 is a two-chain glycosylphosphatidylinositol (GPI)-anchored glycoprotein expressed at multiple stages of B-cell development, beginning with the bone marrow pro-B-cell compartment and continuing through mature, surface Ig positive B-cells. Plasma cell expression is very low or negative. CD24 is also expressed on the majority of B-lineage acute lymphoblastic leukemias, B-cell CCLs and B-cell non-Hodgkin ft.s lymphomas. CD24 may play a role in regulation of B-cell proliferation and maturation, and control of autoimmunity.
Specifications
Specifications
CD24
Monoclonal
FITC
CD24
Liquid
CD24
Human CD24
Purified
ASR
Antibody
Monoclonal
Human
Flow Cytometry
SN3
PBS with 4mg/mL BSA and 0.1% sodium azide
P25063
CD24A, FLJ22950, FLJ43543, MGC75043
Mouse
IgG1
0.5 mL
4° C, store in dark
Primary
100133941