Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
CD24 Mouse anti-Human, PE-Alexa Fluor® 610, Clone: SN3, Invitrogen™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MHCD2422
Catalog No. 50-112-1388
Description
Description
The R-phycoerythrin (PE)-Cy7™ tandem conjugate permits simultaneous multicolor labeling and detection of multiple targets with excitation by a 488 nm laser without occupying traditional channels associated with FITC and PE.
CD24 is a two-chain glycosylphosphatidylinositol (GPI)-anchored glycoprotein expressed at multiple stages of B-cell development, beginning with the bone marrow pro-B-cell compartment and continuing through mature, surface Ig positive B-cells. Plasma cell expression is very low or negative. CD24 is also expressed on the majority of B-lineage acute lymphoblastic leukemias, B-cell CCLs and B-cell non-Hodgkin ft.s lymphomas. CD24 may play a role in regulation of B-cell proliferation and maturation, and control of autoimmunity.
Specifications
Specifications
CD24
Monoclonal
PE-Alexa Fluor™ 610
CD24
Liquid
CD24
Human CD24
Purified
ASR
Antibody
Monoclonal
Human
Flow Cytometry
SN3
PBS with 10% sucrose, BSA and 0.1% sodium azide
P25063
CD24A, FLJ22950, FLJ43543, MGC75043
Mouse
IgG1
0.5 mL
4° C, store in dark
Primary
100133941