Learn More
CD24 Mouse anti-Human, PE, Clone: SN3, Invitrogen™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MHCD2404
Catalog No. 50-112-1387
Add to cart
Description
Description
R-phycoerythrin (PE) is a stable and highly soluble phycobiliprotein which provides maximal absorbance and fluorescence without susceptibility to internal or external fluorescence quenching, thus providing an exceptional quantum yields and molar extinction coefficients.
CD24 is a two-chain glycosylphosphatidylinositol (GPI)-anchored glycoprotein expressed at multiple stages of B-cell development, beginning with the bone marrow pro-B-cell compartment and continuing through mature, surface Ig positive B-cells. Plasma cell expression is very low or negative. CD24 is also expressed on the majority of B-lineage acute lymphoblastic leukemias, B-cell CCLs and B-cell non-Hodgkin ft.s lymphomas. CD24 may play a role in regulation of B-cell proliferation and maturation, and control of autoimmunity.
Specifications
Specifications
CD24
Monoclonal
PE
CD24
Liquid
CD24
Human CD24
Purified
ASR
Antibody
Monoclonal
Human
Flow Cytometry
SN3
PBS with 4mg/mL BSA, sucrose and 0.1% sodium azide
P25063
CD24A, FLJ22950, FLJ43543, MGC75043
Mouse
IgG1
0.5 mL
4° C, store in dark
Primary
100133941