Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD24 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549555

Catalog No. PA549555


Add to cart

Description

Description

CD24 is a two-chain glycosylphosphatidylinositol (GPI)-anchored glycoprotein expressed at multiple stages of B-cell development, beginning with the bone marrow pro-B-cell compartment and continuing through mature, surface Ig positive B-cells. Plasma cell expression is very low or negative. CD24 is also expressed on the majority of B-lineage acute lymphoblastic leukemias, B-cell CCLs and B-cell non-Hodgkin ft.s lymphomas. CD24 may play a role in regulation of B-cell proliferation and maturation, and control of autoimmunity.
Specifications

Specifications

Cd24
Polyclonal
Unconjugated
CD24
Liquid
CD24, Cd24a
synthetic peptide corresponding to 27-53aa of rat CD24
Protein A
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunohistochemistry (Paraffin), Western Blot
5 mg/mL
TBS with 1% BSA, 50% glycerol and 0.03% Proclin 300; pH 7.4
P25063, P24807, Q07490
Signal transducer CD24, Heat-stable antigen, HSA, Nectadrin, CD24, Cd24, Cd24a
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
100133941, 12484, 25145
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit