Learn More
CD24 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 183301AP
Catalog No. 50-173-2117
Add to cart
Description
Description
CD24 is a two-chain glycosylphosphatidylinositol (GPI)-anchored glycoprotein expressed at multiple stages of B-cell development, beginning with the bone marrow pro-B-cell compartment and continuing through mature, surface Ig positive B-cells. Plasma cell expression is very low or negative. CD24 is also expressed on the majority of B-lineage acute lymphoblastic leukemias, B-cell CCLs and B-cell non-Hodgkin ft.s lymphomas. CD24 may play a role in regulation of B-cell proliferation and maturation, and control of autoimmunity.
Specifications
Specifications
CD24
0.18 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P24807, P25063
CD24, CD24 molecule, CD24A, Signal transducer CD24
Rabbit
IgG
150 μL
-20°C
Primary
100133941, 12484
Western Blot
Unconjugated
CD24
Liquid
CD24
CD24 Fusion Protein Ag11679
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse